Merck
  • Acylated ghrelin prevents doxorubicin-induced cardiac intrinsic cell death and fibrosis in rats by restoring IL-6/JAK2/STAT3 signaling pathway and inhibition of STAT1.
Acylated ghrelin prevents doxorubicin-induced cardiac intrinsic cell death and fibrosis in rats by restoring IL-6/JAK2/STAT3 signaling pathway and inhibition of STAT1.
Naunyn-Schmiedeberg's archives of pharmacology (2019-05-17)
Ali A Shati, Attalla Farag El-Kott
ABSTRACT
This study investigated if JAK/STAT signaling pathway mediates doxorubicin (DOX)-induced cell death and fibrosis in left ventricles (LVs) of rats and examined if acylated ghrelin affords protection by modulating this pathway. Male rats (120 ± 5 g) were divided into 6 groups (10 rats each) as follows: control; control + AG (10 ng/kg, s.c.); DOX (an accumulative dose 15 mg/kg, i.p.); DOX + AG, DOX + AG + AG490, a JAK2 inhibitor (5 mg/kg, i.p.); and DOX + AG + [D-Lys3]-GHRP-6; an AG receptor antagonist (3.75 mg/kg, i.p.). All treatments were carried out for 35 days. In rats' LVs, DOX significantly impaired the systolic and diastolic functions, enhanced levels of ROS and MDA, reduced levels of GSH and Bcl-2, and increased mRNA and protein levels of collagen I/III and TGF-β and cleaved caspase-3. In addition, although DOX did not affect JAK1 or JAK2 activity, it significantly increased protein levels of IL-6, decreased STAT3 and p-STAT3 (Tyr701&Ser727), and increased STAT1 and p-STAT1 (Tyr701&Ser727) levels, with a concomitant decrease in ERK1/2 activity and an increase in P38 activity. However, without affecting IL-6 and JAK1/2, AG reversed all of the observed alterations with a significant increase in the levels and activities of JAK2. Similar effects of AG were also seen in control rats. Interestingly, all the beneficial effects afforded by AG were abolished by AG490 and AG + [D-Lys3]-GHRP-6. In conclusion, DOX-induced cardiac toxicity involves stimulation of IL-6, P38, and STAT1 signaling levels whereas the protective effect afforded by AG involves the activation of ERK1/2 and JAK2/STAT3 and inhibition of STAT1.