Merck
  • Inhibition of the Voltage-Dependent K+ Current by the Tricyclic Antidepressant Desipramine in Rabbit Coronary Arterial Smooth Muscle Cells.
Inhibition of the Voltage-Dependent K+ Current by the Tricyclic Antidepressant Desipramine in Rabbit Coronary Arterial Smooth Muscle Cells.
Cardiovascular toxicology (2017-11-15)
Sung Eun Shin, Hongliang Li, Jin Ryeol An, Mi Seon Seo, Sung Hun Na, Won-Kyo Jung, Amy L Firth, Kwon-Soo Ha, Eun-Taek Han, Seok-Ho Hong, Il-Whan Choi, Won Sun Park
ABSTRACT
We describe the effect of a tricyclic antidepressant drug desipramine on voltage-dependent K+ (Kv) currents in freshly isolated rabbit coronary arterial smooth muscle cells using a conventional whole-cell patch clamp technique. Application of desipramine rapidly decreased the Kv current amplitude in a concentration-dependent manner, with an IC50 value of 5.91 ± 0.18 μM and a Hill coefficient of 0.61 ± 0.09. The steady-state inactivation curves of the Kv channels were not affected by desipramine. However, desipramine shifted the steady-state inactivation curves toward a more negative potential. Application of train pulses (1 or 2 Hz) slightly reduced the Kv current amplitude. Such reduction in the Kv current amplitude by train pulses increased in the presence of desipramine. Furthermore, the inactivation recovery time constant was also increased in the presence of desipramine, suggesting that desipramine-induced inhibition of the Kv current was use-dependent. Application of a Kv1.5 inhibitor (DPO-1) and/or a Kv2.1 inhibitor (guangxitoxin) did not change the inhibitory effect of desipramine on Kv currents. Based on these results, we concluded that desipramine directly inhibited the Kv channels in a dose- and state-dependent manner, but the effect was independent of norepinephrine/serotonin reuptake inhibition.