Merck
  • Build/couple/pair strategy combining the Petasis 3-component reaction with Ru-catalyzed ring-closing metathesis and isomerization.
Build/couple/pair strategy combining the Petasis 3-component reaction with Ru-catalyzed ring-closing metathesis and isomerization.
ACS combinatorial science (2012-02-24)
Erhad Ascic, Sebastian T Le Quement, Mette Ishoey, Mathilde Daugaard, Thomas E Nielsen
ABSTRACT
A "build/couple/pair" pathway for the systematic synthesis of structurally diverse small molecules is presented. The Petasis 3-component reaction was used to synthesize anti-amino alcohols displaying pairwise reactive combinations of alkene moieties. Upon treatment with a ruthenium alkylidene-catalyst, these dienes selectively underwent ring-closing metathesis reactions to form 5- and 7-membered heterocycles and cyclic aminals via a tandem isomerization/N-alkyliminium cyclization sequence.
Sigma-Aldrich
trans-2-Phenylvinylboronic acid, 97%