Merck
  • Analysis of peptide secondary structures by photoactivatable amino acid analogues.
Analysis of peptide secondary structures by photoactivatable amino acid analogues.
Angewandte Chemie (International ed. in English) (2012-10-31)
Knut Kölbel, Christian H Ihling, Andrea Sinz
ABSTRACT
Photochemical cross-linking was applied to trap intramolecular interactions in peptides. The incorporation of diazirine-labeled amino acid analogues in combination with high-resolution mass spectrometry made it possible to catch reverse-turn conformations within peptides, exactly map their self-interacting surfaces, and discriminate between stable and transient interactions.
Sigma-Aldrich
H-L-Photo-methionine HCl, ≥95%