Merck
  • p200 RhoGAP promotes cell proliferation by mediating cross-talk between Ras and Rho signaling pathways.
p200 RhoGAP promotes cell proliferation by mediating cross-talk between Ras and Rho signaling pathways.
The Journal of biological chemistry (2007-02-03)
Xun Shang, Sun Young Moon, Yi Zheng
ABSTRACT
p200 RhoGAP, a member of the Rho GTPase-activating protein (RhoGAP) family, was previously implicated in the regulation of neurite outgrowth through its RhoGAP activity. Here we show that ectopic expression of p200 RhoGAP stimulates fibroblast cell proliferation and cell cycle progression, leading to transformation. The morphology of the foci induced by p200 RhoGAP is distinct from that formed by Rac or Rho activation but similar to that induced by oncogenic Ras, raising the possibility that p200 RhoGAP may engage Ras signaling. Expression of p200 RhoGAP results in a significant increase of Ras-GTP and the activation of two downstream signaling pathways of Ras, ERK1/2 and phosphatidylinositol 3-kinase. Inhibition of Ras or ERK1/2, but not phosphatidylinositol 3-kinase, effectively suppresses the foci formation induced by p200 RhoGAP, suggesting that the Ras-ERK pathway is required for p200 RhoGAP-mediated cell transformation. p200 RhoGAP co-localizes with p120 RasGAP in cells and forms a complex with p120 RasGAP, and this interaction is mediated by the C-terminal region and the Src homology 3 domain of p200 RhoGAP and p120 RasGAP, respectively. Mutations of p200 RhoGAP that disrupt interaction with p120 RasGAP abolish its Ras activation and cell transforming activities. Interestingly, the RhoGAP activity of the N-terminal RhoGAP domain in p200 RhoGAP is also required for its full transforming activity, and expression of a dominant negative RhoA mutant that blocks RhoA cycling between the GDP- and GTP-bound states suppresses p200 RhoGAP transformation. These results suggest that a Rho GTPase-activating protein may have a positive input to cell proliferation and provide evidence that p200 RhoGAP can mediate cross-talks between Ras- and Rho-regulated signaling pathways in cell growth regulation.