Merck
  • [Inhibitory effects of BIO-1211 on bronchoconstriction and neutrophil adhesion in rats].
[Inhibitory effects of BIO-1211 on bronchoconstriction and neutrophil adhesion in rats].
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences (2008-08-16)
Xin-wei Dong, Xiao-gang Du, Shui-juan Zhang, Ji-qiang Chen, Qiang-min Xie
ABSTRACT
To determine the inhibitory effects of BIO-1211, a very late antigen-4 (vla-4) antagonist, on bronchoconstriction and neutrophil adhesion in rats. For evaluating ovalbumin-induced bronchoconstriction in the sensitized rats, the changes in lung resistance (RL) and lung dynamic compliance (C(dyn)) were determined after antigen challenge. Neutrophils from the rats were used to determine fibronectin and serum-induced cell adhesion. The effect of BIO-1211 on wheezing was determined after inhalation of histamine and acetylcholine in guinea pigs. BIO-1211 aerosol at 1, 3 and 10 mg/ml significantly inhibited the changes in lung resistance and lung dynamic compliance after antigen challenge in the sensitized rats in a dose-dependent manner. BIO-1211 at 25, 50, 100 and 200 microgram/ml inhibited the fibronectin-induced neutrophil adhesion by 23.5%, 24.6%, 61.4% and 58.1%, respectively, and serum-induced adhesion by 29.9%, 35.9%, 35.3% and 15.4%, respectively. Inhalation of 10 mg/ml BIO-1211 did not show any protection against histamine and acetylcholine-induced bronchoconstriction. BIO-1211 inhibits bronchoconstriction and neutrophil adhesion, which may be associated with its effect against bronchoconstriction in rats.
Sigma-Aldrich
BIO 1211 trifluoroacetate salt, ≥97% (HPLC)