Merck
  • Gene array profile identifies collagen type XV as a novel human osteoblast-secreted matrix protein.
Gene array profile identifies collagen type XV as a novel human osteoblast-secreted matrix protein.
Journal of cellular physiology (2009-04-15)
Gina Lisignoli, Katia Codeluppi, Katia Todoerti, Cristina Manferdini, Anna Piacentini, Nicoletta Zini, Francesco Grassi, Luca Cattini, Roberta Piva, Vittorio Rizzoli, Andrea Facchini, Nicola Giuliani, Antonino Neri
ABSTRACT
Bone marrow stromal cells (MSCs) and osteoblasts are the two main non-haematopoietic cellular components of human bone tissue. To identify novel osteoblast-related molecules, we performed a gene expression profiling analysis comparing MSCs and osteoblasts isolated from the same donors. Genes differentially overexpressed in osteoblasts were mainly related to the negative control of cell proliferation, pro-apoptotic processes, protein metabolism and bone remodelling. Notably, we also identified the collagen XV (COL15A1) gene as the most up-regulated gene in osteoblasts compared with MSCs, previously described as being expressed in the basement membrane in other cell types. The expression of collagen type XV was confirmed at the protein level on isolated osteoblasts and we demonstrated that it significantly increases during the osteogenic differentiation of MSCs in vitro and that free ionised extracellular calcium significantly down-modulates its expression. Moreover, light and electron microscopy showed that collagen type XV is expressed in bone tissue biopsies mainly by working osteoblasts forming new bone tissue or lining bone trabeculae. To our knowledge, these data represent the first evidence of the expression of collagen type XV in human osteoblasts, a calcium-regulated protein which correlates to a specific functional state of these cells.
Sigma-Aldrich
Anti-PECAM-1 Antibody, domains 3-6 of human PECAM-1, clone HC1/6, clone HC1/6, Chemicon®, from mouse