Merck
  • The impact of vascular endothelial growth factor and basic fibroblast growth factor on cardiac fibroblasts grown under altered gravity conditions.
The impact of vascular endothelial growth factor and basic fibroblast growth factor on cardiac fibroblasts grown under altered gravity conditions.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2011-01-12)
Claudia Ulbrich, Annekatrin Leder, Jessica Pietsch, Burkhard Flick, Markus Wehland, Daniela Grimm
ABSTRACT
Myocardium is very sensitive to gravitational changes. During a spaceflight cardiovascular atrophy paired with rhythm problems and orthostatic intolerance can occur. The aim of this study was to investigate the impact of basic fibroblast growth factor (bFGF) and vascular endothelial growth factor (VEGF) on cardiac fibroblasts (CF) grown under altered gravity conditions. We examined the influence of exposure to a Random Positioning Machine (RPM) on CF, derived from porcine hearts. We focused on growth, extracellular matrix protein (ECMP) synthesis and apoptosis. When cultured on a RPM, CF began to form 3D spheroids within 24h, irrespective of growth factor treatment. Exposure to RPM induced an increased synthesis of ECMP and also resulted in elevated apoptosis in adherent CF as measured by terminal deoxynucleotidyl transferase-mediated dUTP digoxigenin nick end labeling (TUNEL) analysis, 4',6-diamidino-2-phenylindole (DAPI) staining, and caspase-3 detection. bFGF and VEGF significantly decreased the amount of ECMP (collagen type I, III, chondroitin sulfate) in 1g and RPM cultures, and also significantly reduced the amount of apoptotic CF as well as caspase-3. Altered gravity conditions on a RPM induced 3D growth, elevated ECMP synthesis and apoptosis in cardiac fibroblasts. Growth factor treatment attenuated programmed cell death and ECMP secretion.
Sigma-Aldrich
2′-Deoxyuridine 5′-triphosphate sodium salt, ≥95%