Merck
  • Functionalized Silver Nano-Sensor for Colorimetric Detection of Hg2+ Ions: Facile Synthesis and Docking Studies.
Functionalized Silver Nano-Sensor for Colorimetric Detection of Hg2+ Ions: Facile Synthesis and Docking Studies.
Sensors (Basel, Switzerland) (2018-08-18)
Kollur Shiva Prasad, Govindaraju Shruthi, Chandan Shivamallu
ABSTRACT
In the present study, we describe the facile synthesis of silver nanoparticles (AgNPs) and their nanostructures functionalized with 2-aminopyrimidine-4,6-diol (APD-AgNPs) for Hg2+ ion detection. The promising colorimetric response of APD-AgNPs to detect Hg2+ ions was visible with naked eyes and spectroscopic changes were examined by using a UV-Visible spectrophotometer. The aggregation of APD-AgNPs upon addition of Hg2+ ions was due to the chelation effect of the functionalized nanostructures and results in a color change from pale brown to deep yellow color. The probing sensitivity was observed within five minutes with a detection limit of about 0.35 µM/L. The TEM images of APD-AgNPs showed polydispersed morphologies with hexagonal, heptagonal and spherical nanostructures with an average size between 10 to 40 nm. Furthermore, the sensing behavior of APD-AgNPs towards Hg2+ ions detection was investigated using docking and interaction studies.
Sigma-Aldrich
2-Amino-4,6-dihydroxypyrimidine, 98%