
Zolmitriptan impurity A

Synonym(s):
(R)-4-({3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl}methyl)oxazolidin-2-one
Empirical Formula (Hill Notation):
C16H21N3O2
CAS Number:
Molecular Weight:
287.36
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing