Skip to Content
SK
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Spectrafuge
™
mini-centrifuge
Products
Product Category
centrifuges (5)
Brand
Sigma-Aldrich (4)
Color
Manufacturer
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
S7816
Spectrafuge
™
mini-centrifuge,
blue, AC/DC input 230 V AC
Expand
View Pricing
S6941
Spectrafuge
™
mini-centrifuge,
red, AC/DC input 230 V AC
Expand
View Pricing
S6816
Spectrafuge
™
mini-centrifuge,
teal, AC/DC input 230 V AC
Expand
View Pricing
S7441
Spectrafuge
™
mini-centrifuge,
gray, AC/DC input 230 V AC
Expand
View Pricing
S7316
Spectrafuge
™
mini-centrifuge,
purple, AC/DC input 230 V AC
Expand
View Pricing