
Pilocarpine nitrate for system suitability

Synonym(s):
Pilocarpine nitrate salt
Empirical Formula (Hill Notation):
C11H16N2O2 · HNO3
CAS Number:
Molecular Weight:
271.27
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing