Skip to Content
Merck
N-(n-Butyl)thiophosphoric triamide analytical standard

N-(n-Butyl)thiophosphoric triamide

N-(n-Butyl)thiophosphoric triamide analytical standard
Synonym(s):
N-Butylphosphorothioic triamide, N-Butylthiophosphoric triamide
Empirical Formula (Hill Notation):
C4H14N3PS
CAS Number:
Molecular Weight:
167.21

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing