Skip to Content
Merck
Neryl acetate analytical standard

Neryl acetate

Neryl acetate analytical standard
Synonym(s):
cis-3,7-Dimethyl-2,6-octadien-1-yl acetate
Linear Formula:
CH3CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Format
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing