Skip to Content
Merck
vibrant-m

Methyl 4-acetamido-2-hydroxybenzoate

vibrant-m
Synonym(s):
Methyl 4-(acetylamino)salicylate, Methyl 4-acetamido-2-hydroxybenzoate
Empirical Formula (Hill Notation):
C10H11NO4
CAS Number:
Molecular Weight:
209.20

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1