Skip to Content
Merck
L-Galactono-1,4-lactone ≥95.0% (GC)

L-Galactono-1,4-lactone

L-Galactono-1,4-lactone ≥95.0% (GC)
Synonym(s):
L-Galactonic acid γ-lactone, NSC 25282
Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing