![Inhibitor Resistant Genotyping PCR ReadyMix™ for Bio-Rad, Cepheid, Eppendorf, Illumina, Corbett, and Roche systems](/deepweb/assets/sigmaaldrich/product/images/210/077/76cda0f2-4a16-4a83-a3c5-1e8139a97b17/640/76cda0f2-4a16-4a83-a3c5-1e8139a97b17.jpg)
Inhibitor Resistant Genotyping PCR ReadyMix™
![Inhibitor Resistant Genotyping PCR ReadyMix™ for Bio-Rad, Cepheid, Eppendorf, Illumina, Corbett, and Roche systems](/deepweb/assets/sigmaaldrich/product/images/210/077/76cda0f2-4a16-4a83-a3c5-1e8139a97b17/640/76cda0f2-4a16-4a83-a3c5-1e8139a97b17.jpg)
Synonym(s):
Inhibitor Resistant Genotyping PCR ReadyMix™
Products
Product Category
Brand
Color
Physical Form
Number of Reactions
Quality Segment
dNTPs
Hot Start
PCR Application
Sample Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing