![Fensulfothion PO-sulfone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/241/929/ec80316d-621a-467a-b45e-9e1e259cf0a4/640/ec80316d-621a-467a-b45e-9e1e259cf0a4.png)
Fensulfothion PO-sulfone
![Fensulfothion PO-sulfone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/241/929/ec80316d-621a-467a-b45e-9e1e259cf0a4/640/ec80316d-621a-467a-b45e-9e1e259cf0a4.png)
Synonym(s):
Diethyl 4-(methylsulfonyl)phenyl phosphate
Empirical Formula (Hill Notation):
C11H17O6PS
CAS Number:
Molecular Weight:
308.29
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing