
Clobetasone butyrate for system suitability

Synonym(s):
Clobetasone butyrate, 21-Chloro-9-fluoro-17-hydroxy-16-methylpregna-1,4-diene-3,11,20-trione 17-butyrate
Empirical Formula (Hill Notation):
C26H32ClFO5
CAS Number:
Molecular Weight:
478.98
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing