![cis-11-Eicosenoic acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/218/313/232159c7-cd07-410a-be58-41048fd326cf/640/232159c7-cd07-410a-be58-41048fd326cf.png)
cis-11-Eicosenoic acid
![cis-11-Eicosenoic acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/218/313/232159c7-cd07-410a-be58-41048fd326cf/640/232159c7-cd07-410a-be58-41048fd326cf.png)
Synonym(s):
Gondoic acid
Linear Formula:
CH3(CH2)7CH=CH(CH2)9CO2H
CAS Number:
Molecular Weight:
310.51
Products
Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Tissue
Quality Segment
Functional Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing