Skip to Content
Merck
Cardiolipin disodium salt from bovine heart ≥97% (TLC), 5 mg/mL in ethanol

Cardiolipin disodium salt from bovine heart

Cardiolipin disodium salt from bovine heart ≥97% (TLC), 5 mg/mL in ethanol
Synonym(s):
1,3-Di(3-sn-phosphatidyl)-sn-glycerol disodium salt
EC Number:

Products

Product Category
Brand
Biological Source
Physical Form
Purity
Tissue
Chemical Composition
Showing 1-1 of 1
Compare
Product Number
Description
Pricing