Skip to Content
Merck
Boc-Lys-OH 99%

Boc-Lys-OH

Boc-Lys-OH 99%
Synonym(s):
Nα-(tert-Butoxycarbonyl)-L-lysine, Nα-Boc-L-lysine, Boc-Lys-OH
Linear Formula:
NH2(CH2)4CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
246.30

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing