![4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/139/662/1ce3ff3b-f928-4434-88d9-f4981db0fc47/640/1ce3ff3b-f928-4434-88d9-f4981db0fc47.png)
4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
![4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/139/662/1ce3ff3b-f928-4434-88d9-f4981db0fc47/640/1ce3ff3b-f928-4434-88d9-f4981db0fc47.png)
Empirical Formula (Hill Notation):
C15H14ClNO2
CAS Number:
Molecular Weight:
275.73
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing