2,3-Diphospho-D-glyceric acid penta(cyclohexylammonium) salt
Synonym(s):
D-Glycerate 2,3-diphosphate pentacyclohexylamine salt
Empirical Formula (Hill Notation):
C3H8O10P2 · 5C6H13N
CAS Number:
Molecular Weight:
761.91
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Tissue
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing