Product Name
9-Chloro-9-phenylxanthene 97%
Product Number
218855
Product Brand
aldrich
CAS Number
42506-03-6
Molecular Weight
292.76
Test | Specification |
---|---|
APPEARANCE (COLOR) | WHITE TO ORANGE TO TAN |
APPEARANCE (FORM) | Powder or Crystals |
TITRATION | 96.5 - 103.5 % |
TITRATION (METHOD) | WITH AGNO3 AFTER O2 COMBUSTION |
PURITY (HPLC AREA %) | ≥ 96.5 % |
SOLUBILITY (COLOR) | Yellow and Orange and Yellow-Orange |
SOLUBILITY (TURBIDITY) | Clear |
SOLUBILITY (METHOD) | 25MG/ML (2.5%) CHLOROFORM |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |