EP5702000515
Eppendorf® Centrifuge 5702
with rotor A-4-38 and 13/16 mm adapters, AC/DC input 230 V (EU plug)
Synonym(s):
Centrifuge
About This Item
Recommended Products
feature
to hold 20 × 15 mL OR 4 × 50 mL conical tubes OR 36 × 13 mm OR 30 × 16 mm blood collection tubes
with rotor A-4-38 and 13/16 mm adapters
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5702000515
parameter
4,400 rpm speed
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service