Merck
  • Coupling process study of lipid production and mercury bioremediation by biomimetic mineralized microalgae.
Coupling process study of lipid production and mercury bioremediation by biomimetic mineralized microalgae.
Bioresource technology (2017-07-15)
Yang Peng, Aosong Deng, Xun Gong, Xiaomin Li, Yang Zhang
ABSTRACT
Considering the high concentration of mercury in industrial wastewater, such as coal-fired power plants and gold mining wastewater, this research study investigated the coupling process of lipid production and mercury bioremediation using microalgae cells. Chlorella vulgaris modified by biomimetic mineralization. The cultivation was divided in two stages: a natural cultivation for 7days and 5days of Hg
Sigma-Aldrich
N,N-Diallylamine hydrochloride, AldrichCPR
Pricing and availability is not currently available.