Merck
  • Role of white adipose lipolysis in the development of NASH induced by methionine- and choline-deficient diet.
Role of white adipose lipolysis in the development of NASH induced by methionine- and choline-deficient diet.
Biochimica et biophysica acta (2014-09-03)
Naoki Tanaka, Shogo Takahashi, Zhong-Ze Fang, Tsutomu Matsubara, Kristopher W Krausz, Aijuan Qu, Frank J Gonzalez
ABSTRACT
Methionine- and choline-deficient diet (MCD) is a model for nonalcoholic steatohepatitis (NASH) in rodents. However, the mechanism of NASH development by dietary methionine/choline deficiency remains undetermined. To elucidate the early metabolic changes associated with MCD-NASH, serum metabolomic analysis was performed using mice treated with MCD and control diet for 3 days and 1 week, revealing significant increases in oleic and linoleic acids after MCD treatment. These increases were correlated with reduced body weight and white adipose tissue (WAT) mass, increased phosphorylation of hormone-sensitive lipase, and up-regulation of genes encoding carboxylesterase 3 and β2-adrenergic receptor in WAT, indicating accelerated lipolysis in adipocytes. The changes in serum fatty acids and WAT by MCD treatment were reversed by methionine supplementation, and similar alterations were detected in mice fed a methionine-deficient diet (MD), thus demonstrating that dietary methionine deficiency enhances lipolysis in WAT. MD treatment decreased glucose and increased fibroblast growth factor 21 in serum, thus exhibiting a similar metabolic phenotype as the fasting response. Comparison between MCD and choline-deficient diet (CD) treatments suggested that the addition of MD-induced metabolic alterations, such as WAT lipolysis, to CD-induced hepatic steatosis promotes liver injury. Collectively, these results demonstrate an important role for dietary methionine deficiency and WAT lipolysis in the development of MCD-NASH.
Product Description
Supelco
Choline Bitartrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Choline bitartrate