Merck
  • Mononuclear metallacyclic silver(I) complexes of third generation bis(1-pyrazolyl)methane ligands.
Mononuclear metallacyclic silver(I) complexes of third generation bis(1-pyrazolyl)methane ligands.
Inorganic chemistry (2009-01-06)
Daniel L Reger, Elizabeth A Foley, Mark D Smith
ABSTRACT
The third-generation bis(1-pyrazolyl)methane ligands p-C(6)H(4)[CH(2)OCH(2)CH(pz)(2)](2) (L(p), pz = pyrazolyl ring) and m-C(6)H(4)[CH(2)OCH(2)CH(pz)(2)](2) (L(m)) have been synthesized by the reaction of (pz)(2)CHCH(2)OH with NaH followed by alpha,alpha'-dibromo-p-xylene or alpha,alpha'-dibromo-m-xylene. The reaction of L(p) with AgBF(4), AgPF(6), and AgO(3)SCF(3) yields the new compounds {Ag[p-C(6)H(4)(CH(2)OCH(2)CH(pz)(2))(2)]}BF(4), {Ag[p-C(6)H(4)(CH(2)OCH(2)CH(pz)(2))(2)]}PF(6), and {Ag[p-C(6)H(4)(CH(2)OCH(2)CH(pz)(2))(2)]}O(3)SCF(3), respectively. A similar reaction of L(m) with AgBF(4) and AgPF(6) yields {Ag[m-C(6)H(4)(CH(2)OCH(2)CH(pz)(2))(2)]}BF(4) and {Ag[m-C(6)H(4)(CH(2)OCH(2)CH(pz)(2))(2)]}PF(6). These compounds were crystallized from both acetone and acetonitrile to yield nine crystalline forms of (LAg)(+) that differ in counterion and solvent of crystallization. In all complexes, the four pyrazolyl rings of the ligand chelate a single silver(I) cation in a distorted tetrahedral environment to form mononuclear metallacycles. This arrangement has not previously been observed with the analogous ligands based on tris(1-pyrazolyl)methane units and is unique because of the ring sizes (16-member rings in L(m) and 17-member rings in L(p)). The dominant feature in all of these solid state structures, regardless of solvent or anion, is this cationic metallacyclic architecture, which does not readily lend itself to strong supramolecular organization.
Sigma-Aldrich
α,α′-Dibromo-p-xylene, purum, ≥98.0% (GC)