Merck
  • HER2-overexpressing breast cancer: FDG uptake after two cycles of chemotherapy predicts the outcome of neoadjuvant treatment.
HER2-overexpressing breast cancer: FDG uptake after two cycles of chemotherapy predicts the outcome of neoadjuvant treatment.
British journal of cancer (2013-08-15)
D Groheux, S Giacchetti, M Hatt, M Marty, L Vercellino, A de Roquancourt, C Cuvier, F Coussy, M Espié, E Hindié
ABSTRACT
Pathologic complete response (pCR) to neoadjuvant treatment (NAT) is associated with improved survival of patients with HER2+ breast cancer. We investigated the ability of interim positron emission tomography (PET) regarding early prediction of pathology outcomes. During 61 months, consecutive patients with locally advanced or large HER2+ breast cancer patients without distant metastases were included. All patients received NAT with four cycles of epirubicin+cyclophosphamide, followed by four cycles of docetaxel+trastuzumab. ¹⁸F-fluorodeoxyglucose (¹⁸F-FDG)-PET/computed tomography (CT) was performed at baseline (PET₁) and after two cycles of chemotherapy (PET₂). Maximum standardised uptake values were measured in the primary tumour as well as in the axillary lymph nodes. The correlation between pathologic response and SUV parameters (SUVmax at PET₁, PET₂ and ΔSUVmax) was examined with the t-test. The predictive performance regarding the identification of non-responders was evaluated using receiver operating characteristics (ROC) analysis. Thirty women were prospectively included and 60 PET/CT examination performed. At baseline, 22 patients had PET+ axilla and in nine of them ¹⁸F-FDG uptake was higher than in the primary tumour. At surgery, 14 patients (47%) showed residual tumour (non-pCR), whereas 16 (53%) reached pCR. Best prediction was obtained when considering the absolute residual SUVmax value at PET₂ (AUC=0.91) vs 0.67 for SUVmax at PET₁ and 0.86 for ΔSUVmax. The risk of non-pCR was 92.3% in patients with any site of residual uptake >3 at PET₂, no matter whether in breast or axilla, vs 11.8% in patients with uptake ≤3 (P=0.0001). The sensitivity, specificity, PPV, NPV and overall accuracy of this cutoff were, respectively: 85.7%, 93.8%, 92.3%, 88.2% and 90%. The level of residual ¹⁸F-FDG uptake after two cycles of chemotherapy predicts residual disease at completion of NAT with chemotherapy+trastuzumab with high accuracy. Because many innovative therapeutic strategies are now available (e.g., addition of a second HER2-directed therapy or an antiangiogenic), early prediction of poor response is critical.
Product Description
Epirubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Epirubicin hydrochloride, ≥90% (HPLC)