Merck
  • Inflammation and disease duration have a cumulative effect on the risk of dysplasia and carcinoma in IBD: a case-control observational study based on registry data.
Inflammation and disease duration have a cumulative effect on the risk of dysplasia and carcinoma in IBD: a case-control observational study based on registry data.
International journal of cancer (2013-06-26)
Urpo Nieminen, Airi Jussila, Stig Nordling, Harri Mustonen, Martti A Färkkilä
ABSTRACT
Patients with long-standing inflammatory bowel disease (IBD) have an increased risk for colorectal carcinoma (CRC). Earlier studies suggest that the severity of inflammation is an independent risk factor for CRC in ulcerative colitis (UC). We investigated the role of histological inflammation as a risk factor for colorectal dysplasia or CRC to better target dysplasia surveillance in IBD. By combining our hospital patient registry and pathology database between 1996 and 2008, we identified 183 IBD patients with dysplasia or CRC. The control group was collected from our registry of IBD patients. Histological severe inflammation was present in 41.4% of patients with dysplasia and in 24.1% of patients with CRC, but in only 4.3% of controls. Severe inflammation had an odds ratio (OR) of 31.8 [95% confidence interval (CI): 15.6-64.9] for dysplasia or carcinoma compared to patients with no inflammation. Among patients with mild to moderate inflammation, the OR was 2.6 (95% CI: 1.6-4.1). Disease duration increased the annual risk for dysplasia or CRC by 4.5%. Coexisting primary sclerosing cholangitis (PSC) did not elevate the risk, whereas use of thiopurines (OR = 0.09, 95% CI: 0.02-0.33) and also 5-aminosalicylic acid (OR 0.17, 95% CI: 0.017-1.01) protected against CRC. As conclusion, degree of inflammation and duration of disease cumulatively increase the risk for dysplasia and CRC. PSC was not identified as a risk factor. We demonstrated that use of thiopurines strongly protects against CRC. These results can be applied to better target dysplasia surveillance in IBD patients.
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Supelco
Mesalamine, Pharmaceutical Secondary Standard; Certified Reference Material