Merck
  • [Preliminary study to evaluate the meaning of GGT fractions analysis in a population of workers previously exposed to asbestos].
[Preliminary study to evaluate the meaning of GGT fractions analysis in a population of workers previously exposed to asbestos].
Giornale italiano di medicina del lavoro ed ergonomia (2013-02-15)
R Foddis, A Bonotti, F Sderci, A Pistelli, E Pantani, M Franzini, A Paolicchi, A Baggiani, A Cristaudo
ABSTRACT
The GGT enzyme, considered for years only as a marker of liver disease and alcohol abuse, has now revealed a risk of death for many causes. Through a molecular exclusion chromatography on FPLC system (Fast Protein Liquid Chromatography), it is possible to discriminate four fractions of GGT, defined according to the molecular weight: big-GGT, medium-GGT, small-GGT and free-GGT. The objective was to study the preventing meaning of GGT fractions for asbestos-related diseases. This study was conducted on 129 workers previously exposed to asbestos, 22 patients affected by Malignant Pleural Mesothelioma and 107 healthy workers. Our data demonstrated a statistical significant correlation between the fraction free-GGT with the presence of MPM, suggesting a possible role for this molecule as a biomarker for MPM diagnosis. However, being a preliminary study, further studies are warranted to confirm our results.
Sigma-Aldrich
γ-Glutamyltranspeptidase from porcine kidney, Type IV, 2.0-6.0 units/mg solid
Sigma-Aldrich
γ-Glutamyltranspeptidase from equine kidney, Type VI, 5-12 units/mg solid