Merck
Characterization of an ecto-ATPase activity in Cryptococcus neoformans.
FEMS yeast research (2005-06-14)
Itallo Collopy Junior, Marcio L Rodrigues, Celuta S Alviano, Luiz R Travassos, José Roberto Meyer-Fernandes
ABSTRACT
Cryptococcus neoformans is the causative agent of pulmonary cryptococcosis and cryptococcal meningoencephalitis, which are major clinical manifestations in immunosuppressed patients. In the present study, a surface ATPase (ecto-ATPase) was identified in C. neoformans yeast cells. Intact yeasts hydrolyzed adenosine-5'-triphosphate (ATP) at a rate of 29.36+/-3.36nmol Pi/hx10(8) cells. In the presence of 5 mM MgCl(2), this activity was enhanced around 70 times, and an apparent K(m) for Mg-ATP corresponding to 0.61mM was determined. Inhibitors of phosphatases, mitochondrial Mg(2+)-ATPases, V-ATPases, Na(+)-ATPases or P-ATPases had no effect on the cryptococcal ATPase, but extracellular impermeant compounds reduced enzyme activity in living cells. ATP was the best substrate for the cryptococcal ecto-enzyme, but it also efficiently hydrolyzed inosine 5'-triphosphate (ITP), cytidine 5'-triphosphate (CTP), guanosine 5'-triphosphate (GTP) and uridine-5'-triphosphate (UTP). In the presence of ATP, C. neoformans became less susceptible to the antifungal action of fluconazole. Our results are indicative of the occurrence of a C. neoformans ecto-ATPase that may have a role in fungal physiology.
Sigma-Aldrich
Inosine 5′-triphosphate trisodium salt, ≥95%