Merck
  • Elevated platelet microparticle levels in nonvalvular atrial fibrillation: relationship to p-selectin and antithrombotic therapy.
Elevated platelet microparticle levels in nonvalvular atrial fibrillation: relationship to p-selectin and antithrombotic therapy.
Chest (2007-03-16)
Anirban Choudhury, Irene Chung, Andrew D Blann, Gregory Y H Lip
ABSTRACT
Platelet microparticles (PMPs), are procoagulant membrane vesicles that are derived from activated platelets, the levels of which are elevated in patients with hypertension, coronary artery disease (CAD), diabetes, and stroke, all of which are conditions that lead to (and are associated with) atrial fibrillation (AF). We hypothesized the following: (1) PMP levels are elevated in patients with AF compared to levels in both healthy control subjects (ie, patients without cardiovascular diseases who are in sinus rhythm) and disease control subjects (ie, patients with hypertension, CAD, diabetes or stroke, but who are in sinus rhythm); (2) PMP levels correlate with levels of soluble P-selectin (sP-selectin) [a marker of platelet activation]; and (3) PMP levels are related to the underlying factors in patients with AF that contribute to the overall risk of stroke secondary to AF. We performed a case-control study of 70 AF patients, 46 disease control subjects and 33 healthy control subjects. Peripheral venous levels of PMP and sP-selectin were analyzed by flow cytometry and enzyme-linked immunosorbent assay, respectively. Both AF patients and disease control subjects had significantly higher levels of PMPs (p < 0.001) and sP-selectin (p = 0.001) compared to healthy control subjects, but there was no difference between AF patients and disease control subjects. There was no difference in PMP levels between patients with paroxysmal and permanent AF (p = 0.581), and between those receiving therapy with aspirin and warfarin (p = 0.779). No significant correlation was observed between PMP and sP-selectin levels (p = 0.463), and the clinical characteristics that contribute to increased stroke risk in patients with AF. On stepwise multiple regression analysis in the combined cohort of AF patients plus disease control subjects, the presence/absence of AF was not an independent determinant of PMP and sP-selectin levels. There is evidence of platelet activation (ie, high PMP and sP-selectin levels) in AF patients, but this is likely to be due to underlying cardiovascular diseases rather than the arrhythmia per se.
Sigma-Aldrich
Latex beads, amine-modified polystyrene, fluorescent red, aqueous suspension, 1.0 μm mean particle size