
Vigabatrin impurity E

Synonym(s):
2-[(2RS)-2-Aminobut-3-enyl]propanedioic acid
Empirical Formula (Hill Notation):
C7H11NO4
CAS Number:
Molecular Weight:
173.17
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing