Skip to Content
Merck
S-(−)-Carbidopa ≥98% (TLC), powder

S-(−)-Carbidopa

S-(−)-Carbidopa ≥98% (TLC), powder
Synonym(s):
(S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoic acid, S-(−)-α-Hydrazino-3,4-dihydroxy-2-methylbenzenepropanoic acid
Empirical Formula (Hill Notation):
C10H14N2O4
CAS Number:
Molecular Weight:
226.23
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing