
Perindopril Related Compound A

Synonym(s):
L-Octahydroindole-2-carboxylic acid, (2S,3aS,7aS)-Octahydro-1H-indole-2-carboxylic acid
Empirical Formula (Hill Notation):
C9H15NO2
CAS Number:
Molecular Weight:
169.22
Products
Brand
Formula Weight
Melting Point (°C)
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing