
Pefabloc® SC

Synonym(s):
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, AEBSF
Empirical Formula (Hill Notation):
C8H10FNO2S · HCl
CAS Number:
Molecular Weight:
239.69
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing