Skip to Content
Merck
Neoeriocitrin ~95% (HPLC)

Neoeriocitrin

Neoeriocitrin ~95% (HPLC)
Synonym(s):
(S)-3′,4′,5,7-Tetrahydroxyflavanone-7-[2-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside], Eriodictyol 7-neohesperidoside
Empirical Formula (Hill Notation):
C27H32O15
CAS Number:
Molecular Weight:
596.53
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing