
Monoclonal Anti-Biotin−Peroxidase antibody produced in mouse

Synonym(s):
Monoclonal Anti-Biotin
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Conjugate
Quality Segment
Package Size
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing