
(−)-Menthone

Synonym(s):
(1R,4S)-p-Menthan-3-one, (2S,5R)-2-Isopropyl-5-methylcyclohexanone
Empirical Formula (Hill Notation):
C10H18O
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4