
Doxazosin Related Compound C

Synonym(s):
4-Amino-2-chloro-6,7-dimethoxyquinazoline, 2-Chloro-6,7-dimethoxy-4-quinazolinamine
Empirical Formula (Hill Notation):
C10H10ClN3O2
CAS Number:
Molecular Weight:
239.66
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2