
Cytochalasin B from Drechslera dematioidea

Synonym(s):
Phomin
Empirical Formula (Hill Notation):
C29H37NO5
CAS Number:
Molecular Weight:
479.61
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-2 of 2
Compare
Product Number
Description
Pricing