Skip to Content
Merck
Cyclophosphamide Related Compound B certified reference material, pharmaceutical secondary standard

Cyclophosphamide Related Compound B

Cyclophosphamide Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
3-(2-Chloroethyl)-2-oxo-2-hydroxy-1,3,6,2-oxadiazaphosphonane
Empirical Formula (Hill Notation):
C7H16ClN2O3P
CAS Number:
Molecular Weight:
242.64

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade