
1-Methyl-1,2,3,4-tetrahydro-β-carboline-3-carboxylic acid

Synonym(s):
1,2,3,4-Tetrahydroharmane-3-carboxylic acid, 1-Methyl-1,2,3,4-Tetrahydropyrido[3,4-b]indole-3-carboxylic acid, 1-Methyl-1,2,3,4-tetrahydro-β-carboline-3-carboxylic acid, NSC 26225
Empirical Formula (Hill Notation):
C13H14N2O2
CAS Number:
Molecular Weight:
230.26
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing