- Inhibition of α5 subunit-containing GABA
Inhibition of α5 subunit-containing GABA
European journal of pain (London, England) (2017-02-02)
M Xue, J P Liu, Y H Yang, Z W Suo, X Yang, X D Hu
PMID28146318
摘要
γ-Aminobutyric acid (GABA) type A receptors (GABA The C fibre-evoked field potentials were recorded in superficial dorsal horn of spinal cord, and the effects of α5-GABA Inhibition of α5-GABA α5-GABA Tonic inhibition generated by α5-GABA
材料