跳转至内容
Merck
  • Inflammation-Related Tumor Progression in Murine Fibrosarcoma Exhibited Over-expression of Sex-determining Region Y-box 2 (Sox2) Compared to Parental Regressor Cells.
Inflammation-Related Tumor Progression in Murine Fibrosarcoma Exhibited Over-expression of Sex-determining Region Y-box 2 (Sox2) Compared to Parental Regressor Cells.
Anticancer research (2015-05-31)
Yasuhiro Kuramitsu, Issei Tanaka, Yufeng Wang, Futoshi Okada, Kazuhiro Tokuda, Takao Kitagawa, Junko Akada, Kazuyuki Nakamura
摘要
Tumor progression is one of the most serious issues to overcome cancer disease. As a model of inflammation-induced tumor progression, we used the regressive murine fibrosarcoma cell clone QR-32 and the progressive malignant clone QRsP-11, that was derived from QR-32. Heat shock protein beta-1 (Hspb1) is a molecular chaperone. Hspb1 plays roles in not only cell protection but also chemo-resistance, tumorigenicity and protection from apoptosis. In a recent study, we showed that Hspb1 was up-regulated in QRsP-11 compared to QR-32. We compared the expression levels of Hspb1, Hsf1 and Sox2 in QR-32 and QRsP-11 cells by means of western blotting. Hsf1, a transcription factor for Hspb1 was not increased in QRsP-11. Sex determining region Y-box 2 (Sox2) is a transcription factor, reported to interact with Hspb1. Sox2 was up-regulated in QRsP-11 compared to QR-32. These results suggest that Sox2-Hspb1 signaling is a possible pathway responsible to tumor progression of QRsP-11.