跳转至内容
Merck
  • Bilobalide attenuates hypoxia induced oxidative stress, inflammation, and mitochondrial dysfunctions in 3T3-L1 adipocytes via its antioxidant potential.
Bilobalide attenuates hypoxia induced oxidative stress, inflammation, and mitochondrial dysfunctions in 3T3-L1 adipocytes via its antioxidant potential.
Free radical research (2014-07-22)
A Priyanka, V M Nisha, S S Anusree, K G Raghu
摘要
Excessive expansion of white adipose tissue leads to hypoxia which is considered as a key factor responsible for adipose tissue dysfunction in obesity. Hypoxia induces inflammation, insulin resistance, and other obesity related complications. So the hypoxia-signalling pathway is expected to provide a new target for the treatment of obesity-associated complications. Inhibition or downregulation of the HIF-1 pathway could be an effective target for the treatment of obesity related hypoxia. In the present study, we evaluated the effect of hypoxia on functions of 3T3-L1 adipocytes emphasising on oxidative stress, antioxidant status, inflammation and mitochondrial functions. We have also evaluated the protective role of bilobalide, a bioactive from Gingko biloba, on hypoxia induced alterations. The results revealed that hypoxia significantly altered all the vital parameters of adipocyte biology like HIF-1α expression (103.47% ↑), lactate and glycerol release (184.34% and 69.1% ↑, respectively), reactive oxygen species (ROS) production (432.53% ↑), lipid and protein oxidation (376.6% and 566.6% ↑, respectively), reduction in antioxidant enzymes (superoxide dismutase and catalase) status, secretion of inflammatory markers (TNF-α, IL-6, IL-1β and IFN-γ) and mitochondrial functions (mitochondrial mass, membrane potential, permeability transition pore integrity, superoxide generation). Bilobalide significantly protected adipocytes from adverse effects of hypoxia in a dose-dependent manner by attenuating oxidative stress, inflammation and protecting mitochondria. Acriflavine (HIF-1 inhibitor) was used as positive control. On the basis of this study, a detailed investigation is needed to delineate the mechanism of action of bilobalide to develop it as therapeutic target for obesity.
材料
货号
品牌
产品描述
Sigma-Aldrich
二甲基亚砜, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
二甲基亚砜, ACS reagent, ≥99.9%
Sigma-Aldrich
二甲基亚砜, for molecular biology
Sigma-Aldrich
二甲基亚砜, suitable for HPLC, ≥99.7%
Sigma-Aldrich
甘油, ACS reagent, ≥99.5%
Sigma-Aldrich
二甲基亚砜, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
甘油, for molecular biology, ≥99.0%
Sigma-Aldrich
二甲基亚砜, ReagentPlus®, ≥99.5%
Sigma-Aldrich
甘油, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
二甲基亚砜, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
地塞米松, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
3-异丁基-1-甲基黄嘌呤, ≥99% (HPLC), powder
Sigma-Aldrich
二甲基亚砜, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
二氯化钴, 97%
Sigma-Aldrich
2',7'-二氯荧光素二乙酸酯, ≥97%
Sigma-Aldrich
3-异丁基-1-甲基黄嘌呤, BioUltra, ≥99%
Sigma-Aldrich
地塞米松, ≥98% (HPLC), powder
Sigma-Aldrich
甘油 溶液, 83.5-89.5% (T)
Sigma-Aldrich
甘油, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
二甲基亚砜, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
甘油, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
甘油, puriss., anhydrous, 99.0-101.0% (alkalimetric)
USP
甘油, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
二氯化钴, purum p.a., anhydrous, ≥98.0% (KT)
Sigma-Aldrich
甘油, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
甘油, FCC, FG
Sigma-Aldrich
二甲基亚砜, PCR Reagent
Sigma-Aldrich
钙黄绿素-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
地塞米松, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
二甲基亚砜, puriss. p.a., dried, ≤0.02% water