跳转至内容
Merck
  • Regional localization of the human G protein alpha i2 (GNAI2) gene: assignment to 3p21 and a related sequence (GNAI2L) to 12p12-p13.
Regional localization of the human G protein alpha i2 (GNAI2) gene: assignment to 3p21 and a related sequence (GNAI2L) to 12p12-p13.
Genomics (1992-01-01)
I Magovcevic, S L Ang, J G Seidman, C J Tolman, E J Neer, C C Morton
摘要
Gi alpha proteins, members of the G protein signal transduction family, include a small number of polypeptides: Gi alpha 1 (GNAI1), Gi alpha 2 (GNAI2), and Gi alpha 3 (GNAI3). A cDNA for the human GNAI2 gene has been isolated from a human T-cell library and is mapped by chromosomal in situ hybridization to the short arm of chromosome 3 at 3p21. A related sequence, GNAI2L, is mapped by in situ hybridization to the short arm of chromosome 12 at p12-p13. These mapping results are further supported by amplification of GNAI2-specific sequences in a monochromosomal human/rodent somatic cell hybrid containing only human chromosome 3. Of note, these assignments are to chromosome regions in which other G proteins reside. Localization of GNAI2 to 3p21 is of great interest as this region of the short arm of chromosome 3 is frequently involved in rearrangements in various human tumors.