跳转至内容
Merck
  • Use of 1,2,4-dithiazolidine-3,5-dione (DtsNH) and 3-ethoxy-1,2,4-dithiazoline-5-one (EDITH) for synthesis of phosphorothioate-containing oligodeoxyribonucleotides.
Use of 1,2,4-dithiazolidine-3,5-dione (DtsNH) and 3-ethoxy-1,2,4-dithiazoline-5-one (EDITH) for synthesis of phosphorothioate-containing oligodeoxyribonucleotides.
Nucleic acids research (1996-05-01)
Q Xu, K Musier-Forsyth, R P Hammer, G Barany
摘要
Previous methods for the preparation of phosphorothioate-containing oligodeoxyribonucleotides rely on the reaction of phosphite triesters with sulfurizing reagents such as tetraethylthiuram disulfide (TETD) and 3H-1,2-benzodithiol-3-one 1,1-dioxide (Beaucage reagent). However, these and other sulfurizing reagents suffer from several disadvantages, and there is great impetus for the development of improved methods for sulfur transfer that are fully compatible with standard automated DNA synthesis. The present report describes the use of 1,2,4-dithiazolidine-3,5-dione (DtsNH) and 3-ethoxy-1,2,4-dithiazoline-5-one (EDITH) as effective sulfurizing reagents that meet these needs. Both reagents are easily prepared, and are stable upon prolonged room temperature storage in acetonitrile solution. The reagents are used at low concentrations (0.05 M) and for short reaction times (30 s). The methodology has been proven for the automated synthesis on 0.2-1.0 micromol scales of oligodeoxyribonucleotides, of length 6-20 bases, containing the phosphorothioate substitution at either a single site or at all positions.
材料
货号
品牌
产品描述
Sigma-Aldrich
3H-1,2-苯并二硫醇-3-酮, 97%